(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid 98+%

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR0574-25GEA 0 DKK
APOSOR0574-25G APOSOR0574-5G APOSOR0574-1G
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid 98+%
(5-Mercapto-1,3,4-thiadiazol-2-ylthio)acetic acid
Formel: C₄H₄N₂O₂S₃
Molekylvægt: 208,28 g/mol
Smeltepunkt: 158…166 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00129970
CAS-nummer: 53723-88-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR