Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III)

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering


Advarsel

APOSIN4839-10GEA 2140 DKK
APOSIN4839-10G
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III)
Tris(2,2,6,6-tetramethyl-3,5-heptanedionato-O,O')bismuth(III)
Formel: C₃₃H₅₇BiO₆
Molekylvægt: 758,782 g/mol
Smeltepunkt: 282 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00064763
CAS-nummer: 142617-53-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR