Scandium(III) carbonate hydrate 99,9%

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSIN3124-1GEA 273 DKK
APOSIN3124-1G
Scandium(III) carbonate hydrate 99,9%
Scandium(III) carbonate hydrate
Formel: Sc₂(CO₃)₃·xH₂O
Molekylvægt: 269,94 g/mol (anhy)
Smeltepunkt: >500 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00149955
CAS-nummer: 5809-49-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR