Erbium(III) oxalate decahydrate 99.99%
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSIN1726-25GEA
819
DKK
APOSIN1726-25G
Erbium(III) oxalate decahydrate 99.99%
Erbium(III) oxalate decahydrate
Formel:
C₆Er₂O₁₂·10H₂O Molekylvægt: 778,73 g/mol (598,58 anhy) Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD00150142 CAS-nummer: 30618-31-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...