Barium isopropoxide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSIN1201-10GEA 0 DKK
APOSIN1201-10G
Barium isopropoxide
Barium isopropoxide
Formel: C₆H₁₄BaO₂
Molekylvægt: 255,501 g/mol
Smeltepunkt: 200 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00050486
CAS-nummer: 24363-37-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR