BMS-582949

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSBISN0294-25MGEA 0 DKK
APOSBISN0294-25MG APOSBISN0294-5MG APOSBISN0294-10MG
BMS-582949
BMS-582949

MAPK/ERK Signalling Pathway

Formel: C₂₂H₂₆N₆O₂
Molekylvægt: 406,48 g/mol
Opbevaringstemperatur: Stue temperatur
CAS-nummer: 623152-17-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR