CB-839

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSBISN0118-10MGEA 0 DKK
APOSBISN0118-10MG APOSBISN0118-50MG APOSBISN0118-5MG
CB-839
CB-839

CB-839 is a potent selective glutaminase inhibitor with IC50 of 24 nM for recombinant human GAC.

Formel: C₂₆H₂₄F₃N₇O₃S
Molekylvægt: 571,57 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD28167826
CAS-nummer: 1439399-58-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR