4-Amino-2-sulphanylpyrimidine-5-carboxamide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR311019-1GEA 4050 DKK
APOSOR311019-1G APOSOR311019-500MG
4-Amino-2-sulphanylpyrimidine-5-carboxamide
4-Amino-2-sulphanylpyrimidine-5-carboxamide
Formel: C₅H₆N₄OS
Molekylvægt: 170,192 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22192593
CAS-nummer: 89323-11-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR