1-Cyclobutyl-4-(piperidine-4-carbonyl)piperazine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310942-500MGEA 6440 DKK
APOSOR310942-500MG
1-Cyclobutyl-4-(piperidine-4-carbonyl)piperazine
1-Cyclobutyl-4-(piperidine-4-carbonyl)piperazine
Formel: C₁₄H₂₅N₃O
Molekylvægt: 251,368 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09261268
CAS-nummer: 799559-59-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR