4-(5-Bromothiophen-2-yl)benzoic acid

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310841-1GEA 0 DKK
APOSOR310841-1G
4-(5-Bromothiophen-2-yl)benzoic acid
4-(5-Bromothiophen-2-yl)benzoic acid
Formel: C₁₁H₇BrO₂S
Molekylvægt: 283,141 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20922788
CAS-nummer: 145208-54-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR