2-(5-Bromo-2-methoxyphenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310586-5GEA 0 DKK
APOSOR310586-5G
2-(5-Bromo-2-methoxyphenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
2-(5-Bromo-2-methoxyphenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
Formel: C₁₇H₁₄BBrN₂O
Molekylvægt: 353,021 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20527134
CAS-nummer: 927384-45-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR