(1R)-1-[3-(4-Chlorophenyl)-1,2-oxazol-5-yl]ethan-1-ol

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310508-1GEA 0 DKK
APOSOR310508-1G
(1R)-1-[3-(4-Chlorophenyl)-1,2-oxazol-5-yl]ethan-1-ol
(1R)-1-[3-(4-Chlorophenyl)-1,2-oxazol-5-yl]ethan-1-ol
Formel: C₁₁H₁₀ClNO₂
Molekylvægt: 223,656 g/mol
Smeltepunkt: 55…68 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD27957328
CAS-nummer: 1565845-64-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR