4-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310503-5GEA 0 DKK
APOSOR310503-5G
4-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde
4-(6-Methyl-4,8-dioxo-1,3,6,2-dioxazaborocan-2-yl)benzaldehyde
Formel: C₁₂H₁₂BNO₅
Molekylvægt: 261,038 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11215233
CAS-nummer: 1072960-66-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR