2-(4-Bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310477-5GEA 2600 DKK
APOSOR310477-5G
2-(4-Bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
2-(4-Bromophenyl)-6-methyl-1,3,6,2-dioxazaborocane-4,8-dione
Formel: C₁₁H₁₁BBrNO₄
Molekylvægt: 311,924 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11215230
CAS-nummer: 943552-04-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR