Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310471-1GEA 1670 DKK
APOSOR310471-1G
Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate
Methyl 6-(tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene-2-carboxylate
Formel: C₁₈H₂₁BO₄
Molekylvægt: 312,168 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18783177
CAS-nummer: 736989-93-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR