(2-Iodo-5-methoxyphenyl)boronic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310361-1GEA 786 DKK
APOSOR310361-1G
(2-Iodo-5-methoxyphenyl)boronic acid
(2-Iodo-5-methoxyphenyl)boronic acid
Formel: C₇H₈BIO₃
Molekylvægt: 277,852 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26793796
CAS-nummer: 89694-50-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR