Methyl 2-butyl-1H-1,3-benzodiazole-6-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310305-1GEA 1840 DKK
APOSOR310305-1G
Methyl 2-butyl-1H-1,3-benzodiazole-6-carboxylate
Methyl 2-butyl-1H-1,3-benzodiazole-6-carboxylate
Formel: C₁₃H₁₆N₂O₂
Molekylvægt: 232,278 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26523303
CAS-nummer: 127007-36-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR