2-(3,5-Dibromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310295-5GEA 0 DKK
APOSOR310295-5G APOSOR310295-1G
2-(3,5-Dibromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
2-(3,5-Dibromophenyl)-2,3-dihydro-1H-naphtho[1,8-de][1,3,2]diazaborinine
Formel: C₁₆H₁₁BBr₂N₂
Molekylvægt: 401,891 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD20527133
CAS-nummer: 1098071-09-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR