2-(Hydroxymethyl)-4,6-dimethylpyridine-3-carbonitrile

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310182-500MGEA 2730 DKK
APOSOR310182-500MG
2-(Hydroxymethyl)-4,6-dimethylpyridine-3-carbonitrile
2-(Hydroxymethyl)-4,6-dimethylpyridine-3-carbonitrile
Formel: C₉H₁₀N₂O
Molekylvægt: 162,188 g/mol
Smeltepunkt: 92…93 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22375454
CAS-nummer: 1383468-71-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR