3,5-Diiodo-6-methylpyridin-2-ol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310094-1GEA 0 DKK
APOSOR310094-1G APOSOR310094-500MG
3,5-Diiodo-6-methylpyridin-2-ol
3,5-Diiodo-6-methylpyridin-2-ol
Formel: C₆H₅I₂NO
Molekylvægt: 360,919 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11100951
CAS-nummer: 858851-91-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR