2-(4-Hydroxyphenoxy)-N-(propan-2-yl)propanamide

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR310018-1GEA 0 DKK
APOSOR310018-1G APOSOR310018-500MG
2-(4-Hydroxyphenoxy)-N-(propan-2-yl)propanamide
2-(4-Hydroxyphenoxy)-N-(propan-2-yl)propanamide
Formel: C₁₂H₁₇NO₃
Molekylvægt: 223,268 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11181693
CAS-nummer: 1094370-49-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR