5,7-Dichloro-2-methylquinoline-3-carboxylic acid
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR309456-1GEA
2510
DKK
APOSOR309456-1G
5,7-Dichloro-2-methylquinoline-3-carboxylic acid
5,7-Dichloro-2-methylquinoline-3-carboxylic acid
Formel:
C₁₁H₇Cl₂NO₂ Molekylvægt: 256,085 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD09787759 CAS-nummer: 948293-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...