4-(3,4-Dimethyl-benzyl)-piperidine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR309446-1GEA 0 DKK
APOSOR309446-1G APOSOR309446-250MG
4-(3,4-Dimethyl-benzyl)-piperidine
4-(3,4-Dimethyl-benzyl)-piperidine
Formel: C₁₄H₂₁N
Molekylvægt: 203,323 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD05863588
CAS-nummer: 782504-68-1

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR