5-Chloro-3-[(4-chlorophenyl)methyl]-1,2,4-thiadiazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR309417-1GEA 0 DKK
APOSOR309417-1G
5-Chloro-3-[(4-chlorophenyl)methyl]-1,2,4-thiadiazole
5-Chloro-3-[(4-chlorophenyl)methyl]-1,2,4-thiadiazole
Formel: C₉H₆Cl₂N₂S
Molekylvægt: 245,128 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14707340
CAS-nummer: 90418-16-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR