3-Amino-7-chloro-2-methylquinoline Dihydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR309365-1GEA 4940 DKK
APOSOR309365-1G
3-Amino-7-chloro-2-methylquinoline Dihydrochloride
3-Amino-7-chloro-2-methylquinoline Dihydrochloride
Formel: C₁₀H₉ClN₂·2ClH
Molekylvægt: 265,567 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18909619
CAS-nummer: 1296950-72-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR