4-Chloro-3,6-dibromoquinoline
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR309335-1GEA
3020
DKK
APOSOR309335-1G
4-Chloro-3,6-dibromoquinoline
4-Chloro-3,6-dibromoquinoline
Formel:
C₉H₄Br₂ClN Molekylvægt: 321,396 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD08437002 CAS-nummer: 927801-19-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...