4-Bromo-6-methoxy-2-phenylquinoline
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR309327-1GEA
3200
DKK
APOSOR309327-1G
4-Bromo-6-methoxy-2-phenylquinoline
4-Bromo-6-methoxy-2-phenylquinoline
Formel:
C₁₆H₁₂BrNO Molekylvægt: 314,177 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD13192938 CAS-nummer: 876491-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...