7-Ethylquinoline-3-carboxylic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR309226-1GEA 3880 DKK
APOSOR309226-1G
7-Ethylquinoline-3-carboxylic acid
7-Ethylquinoline-3-carboxylic acid
Formel: C₁₂H₁₁NO₂
Molekylvægt: 201,221 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09787862
CAS-nummer: 948290-70-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR