2-(Propan-2-yl)morpholine

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR308186-1GEA 5410 DKK
APOSOR308186-1G
2-(Propan-2-yl)morpholine
2-(Propan-2-yl)morpholine
Formel: C₇H₁₅NO
Molekylvægt: 129,2 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD14582705
CAS-nummer: 89855-02-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR