(S)-3-Isopropylmorpholine
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR308182-1GEA
1590
DKK
APOSOR308182-1G
APOSOR308182-250MG
(S)-3-Isopropylmorpholine
(S)-3-Isopropylmorpholine
Formel:
C₇H₁₅NO Molekylvægt: 129,2 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD11519112 CAS-nummer: 77897-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...