N-[4-(Dimethylamino)phenyl]maleimide

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR30796-250MGEA 1760 DKK
APOSOR30796-250MG APOSOR30796-1G
N-[4-(Dimethylamino)phenyl]maleimide
N-[4-(Dimethylamino)phenyl]maleimide
Formel: C₁₂H₁₂N₂O₂
Molekylvægt: 216,236 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD00748633
CAS-nummer: 6953-81-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR