tert-Butyl 4-(4-amino-2-chlorophenyl)piperazine-1-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR307791-1GEA 2450 DKK
APOSOR307791-1G
tert-Butyl 4-(4-amino-2-chlorophenyl)piperazine-1-carboxylate
tert-Butyl 4-(4-amino-2-chlorophenyl)piperazine-1-carboxylate
Formel: C₁₅H₂₂ClN₃O₂
Molekylvægt: 311,807 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11519255
CAS-nummer: 193902-81-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR