2-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR307787-1GEA 3880 DKK
APOSOR307787-1G
2-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester
2-[(4-Amino-phenylamino)-methyl]- piperidine-1-carboxylic acid tert-butyl ester
Formel: C₁₇H₂₇N₃O₂
Molekylvægt: 305,415 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD12026441
CAS-nummer: 1159976-36-9

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR