3-Amino-3-(2,4-dichlorophenyl)-propionic acid

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR307577-1GEA 881 DKK
APOSOR307577-1G
3-Amino-3-(2,4-dichlorophenyl)-propionic acid
3-Amino-3-(2,4-dichlorophenyl)-propionic acid
Formel: C₉H₉Cl₂NO₂
Molekylvægt: 234,079 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD02663185
CAS-nummer: 152606-17-2

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR