7-Bromo-2-hydrazino-3-methylquinoline hydrochloride

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR307526-1GEA 3700 DKK
APOSOR307526-1G
7-Bromo-2-hydrazino-3-methylquinoline hydrochloride
7-Bromo-2-hydrazino-3-methylquinoline hydrochloride
Formel: C₁₀H₁₀BrN₃·ClH
Molekylvægt: 288,571 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11505171
CAS-nummer: 1171639-07-8

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR