7-Bromo-3,4-dichloroquinoline

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR307514-1GEA 3700 DKK
APOSOR307514-1G
7-Bromo-3,4-dichloroquinoline
7-Bromo-3,4-dichloroquinoline
Formel: C₉H₄BrCl₂N
Molekylvægt: 276,945 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD13192969
CAS-nummer: 1021435-01-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR