2-Chloro-3-methoxypyridin-4-ol

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR307236-100MGEA 383 DKK
APOSOR307236-100MG
2-Chloro-3-methoxypyridin-4-ol
2-Chloro-3-methoxypyridin-4-ol
Formel: C₆H₆ClNO₂
Molekylvægt: 159,57 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11553129
CAS-nummer: 1227600-20-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR