2-Methoxy-6-(pyrrolidin-3-yloxy)pyridine dihydrochloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR306650-250MGEA 0 DKK
APOSOR306650-250MG
2-Methoxy-6-(pyrrolidin-3-yloxy)pyridine dihydrochloride
2-Methoxy-6-(pyrrolidin-3-yloxy)pyridine dihydrochloride
Formel: C₁₀H₁₄N₂O₂·2ClH
Molekylvægt: 267,152 g/mol
Smeltepunkt: 146…148 °C
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26097155
CAS-nummer: 1713162-92-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR