4-Methyl-2-(pyrrolidin-3-yloxy)pyridine hydrochloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR306575-1GEA 0 DKK
APOSOR306575-1G
4-Methyl-2-(pyrrolidin-3-yloxy)pyridine hydrochloride
4-Methyl-2-(pyrrolidin-3-yloxy)pyridine hydrochloride
Formel: C₁₀H₁₄N₂O·ClH
Molekylvægt: 214,692 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD24371385
CAS-nummer: 1420994-15-5

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR