3-Phenyl-1-(piperidin-4-yl)-1H-pyrazol-5-amine

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR306416-1GEA 0 DKK
APOSOR306416-1G APOSOR306416-500MG APOSOR306416-250MG
3-Phenyl-1-(piperidin-4-yl)-1H-pyrazol-5-amine
3-Phenyl-1-(piperidin-4-yl)-1H-pyrazol-5-amine
Formel: C₁₄H₁₈N₄
Molekylvægt: 242,32 g/mol
Smeltepunkt: 180 °C (dec.)
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD22375746
CAS-nummer: 1389315-07-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR