2-Bromo-1,4,5-trimethyl-1H-imidazole

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305589-1GEA 5770 DKK
APOSOR305589-1G
2-Bromo-1,4,5-trimethyl-1H-imidazole
2-Bromo-1,4,5-trimethyl-1H-imidazole
Formel: C₆H₉BrN₂
Molekylvægt: 189,053 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD26678978
CAS-nummer: 1218910-27-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR