2-Chloroquinoxaline-6-sulphonyl chloride

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

APOSOR305426-1GEA 0 DKK
APOSOR305426-1G APOSOR305426-250MG
2-Chloroquinoxaline-6-sulphonyl chloride
2-Chloroquinoxaline-6-sulphonyl chloride
Formel: C₈H₄Cl₂N₂O₂S
Molekylvægt: 263,101 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD16547807
CAS-nummer: 877078-00-7
UN nummer: 3261
ADR: 8,III

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR