Methyl 2-(bromomethyl)isonicotinate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

APOSOR305422-250MGEA 0 DKK
APOSOR305422-250MG
Methyl 2-(bromomethyl)isonicotinate
Methyl 2-(bromomethyl)isonicotinate
Formel: C₈H₈BrNO₂
Molekylvægt: 230,059 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD10566277
CAS-nummer: 914639-05-7

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR