3-(Pyridin-3-yl)propane-1-thiol
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
APOSOR305417-250MGEA
0
DKK
APOSOR305417-250MG
APOSOR305417-1G
3-(Pyridin-3-yl)propane-1-thiol
3-(Pyridin-3-yl)propane-1-thiol
Formel:
C₈H₁₁NS Molekylvægt: 153,245 g/mol Opbevaringstemperatur: Stue temperatur |
MDL Number:
MFCD19347136 CAS-nummer: 69603-49-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...