5-Iodo-1,3-dihydro-2H-pyrrolo[2,3-b]pyridin-2-one

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305348-250MGEA 0 DKK
APOSOR305348-250MG
5-Iodo-1,3-dihydro-2H-pyrrolo[2,3-b]pyridin-2-one
5-Iodo-1,3-dihydro-2H-pyrrolo[2,3-b]pyridin-2-one
Formel: C₇H₅IN₂O
Molekylvægt: 260,032 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD15529121
CAS-nummer: 1160112-78-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR