tert-Butyl (4-oxopiperidin-1-yl)acetate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305345-250MGEA 1510 DKK
APOSOR305345-250MG
tert-Butyl (4-oxopiperidin-1-yl)acetate
tert-Butyl (4-oxopiperidin-1-yl)acetate
Formel: C₁₁H₁₉NO₃
Molekylvægt: 213,273 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD07371646
CAS-nummer: 149554-03-0

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR