Methyl 5-amino-1-ethyl-1H-pyrrole-2-carboxylate

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305323-250MGEA 0 DKK
APOSOR305323-250MG
Methyl 5-amino-1-ethyl-1H-pyrrole-2-carboxylate
Methyl 5-amino-1-ethyl-1H-pyrrole-2-carboxylate
Formel: C₈H₁₂N₂O₂
Molekylvægt: 168,193 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD18073423
CAS-nummer: 1379227-91-4

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR