7-Chloro-2,3-dihydro-1H-isoindol-1-one
Leverandør: Apollo Scientific
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
APOSOR305301-250MGEA
3990
DKK
APOSOR305301-250MG
7-Chloro-2,3-dihydro-1H-isoindol-1-one
7-Chloro-2,3-dihydro-1H-isoindol-1-one
Formel:
C₈H₆ClNO Molekylvægt: 167,592 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD09701288 CAS-nummer: 658683-16-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...