Methyl 3-oxo-2,3-dihydro-1H-isoindole-4-carboxylate

Leverandør: Apollo Scientific
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305299-250MGEA 2630 DKK
APOSOR305299-250MG
Methyl 3-oxo-2,3-dihydro-1H-isoindole-4-carboxylate
Methyl 3-oxo-2,3-dihydro-1H-isoindole-4-carboxylate
Formel: C₁₀H₉NO₃
Molekylvægt: 191,183 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD11506113
CAS-nummer: 935269-25-3

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR