6,7-Dimethoxy-2,3-dihydro-1H-isoindol-1-one

Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

APOSOR305296-250MGEA 0 DKK
APOSOR305296-250MG APOSOR305296-1G
6,7-Dimethoxy-2,3-dihydro-1H-isoindol-1-one
6,7-Dimethoxy-2,3-dihydro-1H-isoindol-1-one
Formel: C₁₀H₁₁NO₃
Molekylvægt: 193,199 g/mol
Opbevaringstemperatur: Stue temperatur
MDL Number: MFCD09029854
CAS-nummer: 59084-79-6

Bestil nu

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR